Skip to Content
Merck
All Photos(1)

Key Documents

450103

Sigma-Aldrich

Yttrium(III) chloride

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Yttrium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YCl3
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

impurities

≤150.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

721 °C (lit.)

density

2.67 g/mL at 25 °C (lit.)

SMILES string

Cl[Y](Cl)Cl

InChI

1S/3ClH.Y/h3*1H;/q;;;+3/p-3

InChI key

PCMOZDDGXKIOLL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare yttrium alkyl alkoxide complexes under strictly anhydrous conditions.

Features and Benefits

Will react with potassium alkoxotitanates to form precursors to novel titanium-containing ceramics

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evans, W.J. et al.
Organometallics, 15, 1351-1351 (1996)
Acceleration of the substitution of silanes with Grignard reagents by using either LiCl or YCl3/MeLi.
Naoki Hirone et al.
Angewandte Chemie (International ed. in English), 49(42), 7762-7764 (2010-09-08)
S Hirano et al.
Toxicology and applied pharmacology, 121(2), 224-232 (1993-08-01)
Although radioactive yttrium (Y) has been used for medical treatment, little attention has been directed toward the toxicity of Y. We report time-course and dose-related changes in tissue distribution, subcellular localization, clearance, and acute toxicity of iv-injected yttrium chloride (YCl3)
F Zhang et al.
Physical review letters, 101(14), 148101-148101 (2008-10-15)
Negatively charged globular proteins in solution undergo a condensation upon adding trivalent counterions between two critical concentrations C and C, C <C. This reentrant condensation behavior above C is caused by short-ranged electrostatic interactions between multivalent cations and acidic residues
K Marubashi et al.
Toxicology letters, 99(1), 43-51 (1998-11-04)
We investigated pulmonary clearance of yttrium (Y) and acute lung injury following intratracheal instillation (i.t.) of yttrium chloride (YCl3) in saline- or YCl3-pretreated rats (30 days before the second challenge). About 67% of the initial dose of Y remained in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service