Skip to Content
Merck
All Photos(2)

Key Documents

204919

Sigma-Aldrich

Yttrium(III) chloride hexahydrate

99.999% trace metals basis

Synonym(s):

Yttrium trichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YCl3 · 6H2O
CAS Number:
Molecular Weight:
303.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
solid

Quality Level

Assay

99.999% trace metals basis

form

solid

impurities

≤15.0 ppm Trace Rare Earth Analysis

mp

100 °C (dec.) (lit.)

density

2.18 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Y](Cl)Cl

InChI

1S/3ClH.6H2O.Y/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

IINACGXCEZNYTF-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masakazu Kawashita et al.
Journal of materials science. Materials in medicine, 21(6), 1837-1843 (2010-03-17)
Porous Y2O3 microparticles 500 microm in size were obtained, when 1 wt%-ammonium alginate aqueous solution was dropped into 0.5 M-YCl3 aqueous solution by a Pasteur pipette and the resultant gel microparticles were heat-treated at 1100 degrees C. Small pores less
J Srank et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 68(12), 2163-2168 (2010-08-10)
Separation of (90)Y from a parent solution of (90)Sr-(90)Y in 0.5 M nitric acid by two-stage liquid-liquid extraction has been investigated using centrifugal extractors. The extraction agent used was 0.25 M di-(2-ethylhexyl)phosphoric acid (D2EHPA) in n-dodecane. Back-extraction used 5 M
Hongguang Liu et al.
PloS one, 5(3), e9470-e9470 (2010-03-09)
Optical imaging (OI) techniques such as bioluminescence and fluorescence imaging have been widely used to track diseases in a non-invasive manner within living subjects. These techniques generally require bioluminescent and fluorescent probes. Here we demonstrate the feasibility of using radioactive
Yusuke Takahashi et al.
Environmental toxicology and pharmacology, 34(2), 574-578 (2012-08-03)
Yttrium is used in the production of various electronic devices because the alloy it contains enhances or modifies the properties of other elements. In order to study the cytotoxic action of yttrium, the effect of yttrium chloride (YCl(3)) on the
Acceleration of the substitution of silanes with Grignard reagents by using either LiCl or YCl3/MeLi.
Naoki Hirone et al.
Angewandte Chemie (International ed. in English), 49(42), 7762-7764 (2010-09-08)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service