Skip to Content
Merck

44953

Sigma-Aldrich

Iron(II) lactate hydrate

≥98.0% (dried material)

Synonym(s):

Ferrous lactate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
[CH3CH(OH)COO]2Fe · xH2O
CAS Number:
Molecular Weight:
233.99 (anhydrous basis)
Beilstein:
6099658
EC Number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (dried material)

reaction suitability

core: iron
reagent type: catalyst

loss

12-15% loss on drying

SMILES string

O.CC(O)C(=O)O[Fe]OC(=O)C(C)O

InChI

1S/2C3H6O3.Fe.H2O/c2*1-2(4)3(5)6;;/h2*2,4H,1H3,(H,5,6);;1H2/q;;+2;/p-2

InChI key

FDSNKEJXJCZFLE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

  • Reagent in preparation of carbon-based noble-metal-free PEFC cathode catalysts (oxygen reduction) for use in polymer electrolyte fuel cells (PEFCs)
  • Used for formulation of chemoattractant coatings for artificial reef structures

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Kapsokefalou et al.
International journal of food sciences and nutrition, 56(4), 293-302 (2005-08-13)
The objectives of the present study were: to compare the solubility and dialyzability of various iron fortificants (iron pyrophosphate, ferrous bis-glycinate, ferrous gluconate, ferrous lactate, ferrous sulfate) added, in the presence of ascorbic acid, to pasteurized milk samples produced under
M Blanco et al.
Journal of pharmaceutical and biomedical analysis, 16(2), 255-262 (1997-12-31)
Near infrared diffuse reflectance spectroscopy with a fibre-optic probe was used to determine the water content in ferrous lactate dihydrate. Spectra were recorded by immersing the probe in a beaker containing the ferrous lactate sample. Spectral data were processed by
B I Malope et al.
British journal of haematology, 115(1), 84-89 (2001-11-28)
Laboratory tests used in the diagnosis of iron status lack specificity in defining iron deficiency anaemia (IDA) and anaemia of inflammation (AI). The serum transferrin receptor (sTfR) may provide more information in this regard. The iron status of 561 pre-school
L J Anghileri et al.
In vivo (Athens, Greece), 13(1), 13-20 (1999-04-28)
The synergistic effects of iron overload and ethanol on the liver of mice were studied over a period of 46 weeks. The determination of several parameters (iron, calcium, magnesium, alpha-hydroxyproline, lipid peroxidation, hepatomegalic and splenomegalic indexes) showed that ferrous and
I Narama et al.
Toxicologic pathology, 27(3), 318-324 (1999-06-05)
Eosinophilic gastroenterocolitis with peripheral eosinophilia was induced in rats fed a diet containing 2.5% or 5.0% iron lactate for 3 mo. Additional findings consistent with iron overload were also observed. Microscopically, the lesions consisted of eosinophilic infiltrations in the mucosa

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service