Skip to Content
Merck
All Photos(5)

Key Documents

447536

Sigma-Aldrich

2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane

95%

Synonym(s):

Tetramethylethylene chlorophosphite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12ClO2P
CAS Number:
Molecular Weight:
182.59
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.471 (lit.)

bp

81.5-82 °C/13 mmHg (lit.)

density

1.149 g/mL at 25 °C (lit.)

SMILES string

CC1(C)OP(Cl)OC1(C)C

InChI

1S/C6H12ClO2P/c1-5(2)6(3,4)9-10(7)8-5/h1-4H3

InChI key

WGPCXYWWBFBNSS-UHFFFAOYSA-N

Application

2-Chloro-4,4,5,5-tetramethyl-1,3,2-dioxaphospholane (TMDP) can be used:
  • As a reagent for the phosphitylation of alcohols and heteroatomic nucleophiles, resulting in the formation of useful glycosyl donors and ligands.
  • As a phosphitylation reagent to derivatize lignin samples for 31P NMR analysis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo, Y.; Sulikowski, G. A.,
Journal of the American Chemical Society, 120, 1392-1392 (1998)
Kraft lignin-based rigid polyurethane foam
Li Y and Ragauskas AJ
J. Wood Chem. Technol., 32(3), 210-224 (2012)
Characterization of CO2 precipitated Kraft lignin to promote its utilization
Nagy M, et al.
Green Chemistry, 12(1), 31-34 (2010)
Synthesis of complex carbohydrates and glycoconjugates: enzyme-based and programmable one-pot strategies.
K M Koeller et al.
Chemical reviews, 100(12), 4465-4494 (2001-12-26)
Nontemplate-dependent polymerization processes: polyhydroxyalkanoate synthases as a paradigm
Stubbe J, et al.
Annual Review of Biochemistry, 74(22), 433-480 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service