Skip to Content
Merck
All Photos(2)

Key Documents

391220

Sigma-Aldrich

2-Chloro-1,3,2-dioxaphospholane

97%

Synonym(s):

Cyclic ethylene chlorophosphite, Cyclic ethylene phosphorochloridite, Ethylene chlorophosphite, Ethylene glycol chlorophosphite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4ClO2P
CAS Number:
Molecular Weight:
126.48
Beilstein:
506179
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.489 (lit.)

bp

62 °C/20 mmHg (lit.)

density

1.422 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClP1OCCO1

InChI

1S/C2H4ClO2P/c3-6-4-1-2-5-6/h1-2H2

InChI key

OLSFRDLMFAOSIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphitylating agent used in the synthesis of phosphocholines and spirophosphoranes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rec. Trav. Chim., 113, 293-293 (1994)
Xue Wan et al.
Biomolecules, 9(12) (2019-12-11)
Effective valorization of lignin is crucial to achieve a sustainable, economic and competitive biorefinery of lignocellulosic biomass. In this work, an integrated process was proposed based on a concentrated phosphoric acid plus hydrogen peroxide (PHP) pretreatment to simultaneously facilitate cellulose
Tetrahedron Letters, 35, 5783-5783 (1994)
Ilkoo Noh et al.
Journal of controlled release : official journal of the Controlled Release Society, 320, 283-292 (2020-01-27)
Image-guided therapy, combined with imaging and therapeutic action, forms an attractive system because it can induce outstanding effects at focused locations. However, the conventional liposomes-based system cannot figure in therapeutic or imaging roles themselves, thereby causing the disadvantage of their
Fritz Sieber et al.
Phosphorus, sulfur, and silicon and the related elements, 180(3-4), 647-657 (2006-02-24)
Elemental selenium generated by the photobleaching of selenomerocyanine dyes forms conjugates with serum albumin and serum lipoproteins that are toxic to leukemia and selected solid tumor cells but well tolerated by normal CD34-positive hematopoietic stem and progenitor cells. Serum albumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service