Skip to Content
Merck
All Photos(3)

Key Documents

442682

Sigma-Aldrich

Lithium phosphate monobasic

greener alternative

99%

Synonym(s):

Lithium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiH2PO4
CAS Number:
Molecular Weight:
103.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder or crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>100 °C (lit.)

density

2.5 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].OP(O)([O-])=O

InChI

1S/Li.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

SNKMVYBWZDHJHE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium phosphate monobasic is a class of electrolytic materials that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Precursor for LiMX2O7 (M = Fe, V; x =P, As) cathode materials.1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Guembe-García et al.
Polymers, 12(6) (2020-06-04)
We anchored a colourimetric probe, comprising a complex containing copper (Cu(II)) and a dye, to a polymer matrix obtaining film-shaped chemosensors with induced selectivity toward glycine. This sensory material is exploited in the selectivity detection of glycine in complex mixtures
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)
Electrodes with high power and high capacity for rechargeable lithium batteries
Kang K, et al.
Science, 311(5763), 977-980 (2006)
The Li-ion rechargeable battery: a perspective
Goodenough JB and Park K
Journal of the American Chemical Society, 135(4), 1167-1176 (2013)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service