Skip to Content
Merck
All Photos(1)

Key Documents

440817

Sigma-Aldrich

1-[3-(Trimethoxysilyl)propyl]urea

97%

Synonym(s):

γ-Ureidopropyltrimethoxysilane, 3-Ureidopropyltrimethoxysilane, N -[3-(Trimethoxysilyl)propyl]urea, Ureidopropyltrimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3Si(CH2)3NHCONH2
CAS Number:
Molecular Weight:
222.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 20 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

217-250 °C (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CO[Si](CCCNC(N)=O)(OC)OC

InChI

1S/C7H18N2O4Si/c1-11-14(12-2,13-3)6-4-5-9-7(8)10/h4-6H2,1-3H3,(H3,8,9,10)

InChI key

LVACOMKKELLCHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-[3-(Trimethoxysilyl)propyl]urea is a urea based surface modifying agent that is used for forming self-assembled monolayers (SAMs) on a variety of substrates. It is mainly utilized to immobilize the surface atoms by forming a ligand.

Application

1-[3-(Trimethoxysilyl)propyl]urea capped magnetic nanoparticles can be used as catalyst supports for immobilizing the surface enzymes and nanoparticles. It may be used in the silanization of ceria that finds potential applications as medical agents.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina I Giunta et al.
ACS nano (2020-12-12)
Owing to their outstanding catalytic properties, enzymes represent powerful tools for carrying out a wide range of (bio)chemical transformations with high proficiency. In this context, enzymes with high biocatalytic promiscuity are somewhat neglected. Here, we demonstrate that a meticulous modification
Ureido-modified macroporous hollow silica microspheres for recovery of Wilkinson's catalyst in hydrogenated nitrile butadiene rubber
Ai C, et al.
Powder Technology, 318, 501-506 (2017)
Magnetic nanoparticles entrapped in siliceous mesocellular foam: a new catalyst support
Lee SS, et al.
Chemistry?A European Journal , 18(24), 7394-7403 (2012)
Influence of surface charge on lysozyme adsorption to ceria nanoparticles
Wang B, et al.
Applied Surface Science, 258(14), 5332-5341 (2012)
Immobilization of alkaline phosphatase on modified silica coatings
Ehlert N, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 131(1-3), 51-57 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service