Skip to Content
Merck
All Photos(3)

Key Documents

43961

Sigma-Aldrich

1-Allyl-3-methylimidazolium chloride

greener alternative

≥97.0% (HPLC)

Synonym(s):

AMIM-Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11ClN2
CAS Number:
Molecular Weight:
158.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (HPLC)

form

crystals

greener alternative product characteristics

Safer Solvents and Auxiliaries
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

SMILES string

[Cl-].C[n+]1ccn(CC=C)c1

InChI

1S/C7H11N2.ClH/c1-3-4-9-6-5-8(2)7-9;/h3,5-7H,1,4H2,2H3;1H/q+1;/p-1

InChI key

QVRCRKLLQYOIKY-UHFFFAOYSA-M

General description

1-Allyl-3-methylimidazolium chloride is a thermostable and nonvolatile task-specific ionic liquid (TSIL). It shows high cellulose solubilization ability.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

AMIM-Cl may be used:
  • As a solvent to dissolve cellulose during the preparation of regenerated cellulose materials.
  • As a solvent for the acetylation of cornhusk cellulose to form cornhusk cellulose acetates (CCAs).
  • As a plasticizer for cornstarch to form AMIM-Cl-plasticized starch film.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganeswar Sahu et al.
Polymers, 12(2) (2020-02-13)
This paper presents the dielectric characteristics of nanocomposite films of poly(vinyl alcohol) (PVA) embedded with silver (Ag) nanoparticles and graphene oxide(GO). The nanocomposite films were fabricated by using the solvent casting approach. The morphological analysis was carried out through scanning
Acetone-soluble cellulose acetates prepared by one-step homogeneous acetylation of cornhusk cellulose in an ionic liquid 1-allyl-3-methylimidazolium chloride (AmimCl).
Cao Y, et al.
Carbohydrate Polymers, 69(4), 665-672 (2007)
Ashna Rajeev et al.
Soft matter, 14(37), 7615-7624 (2018-08-31)
Water added to a solution of microcrystalline cellulose (MCC) in 1-allyl-3-methylimidazolium chloride (AmimCl) reduces the solvent quality and causes significant changes in the flow properties and microstructure due to restructuring and aggregation of cellulose molecules. We report an experimental investigation
Tosylation and acylation of cellulose in 1-allyl-3-methylimidazolium chloride.
Granstrom M, et al.
Cellulose, 15(3), 481-488 (2008)
1-Allyl-3-methylimidazolium chloride room temperature ionic liquid: a new and powerful nonderivatizing solvent for cellulose.
Zhang H, et al.
Macromolecules, 38(20), 8272-8277 (2005)

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service