Skip to Content
Merck
All Photos(1)

Key Documents

439312

Sigma-Aldrich

1-Fluoro-2,4,6-trimethylpyridinium tetrafluoroborate

≥95%

Synonym(s):

1-Fluoro-sym-collidinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11BF5N
CAS Number:
Molecular Weight:
226.98
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

reaction suitability

reaction type: C-H Activation
reagent type: catalyst
reagent type: oxidant
reaction type: Fluorinations

mp

213-217 °C (lit.)

SMILES string

F[B-](F)(F)F.Cc1cc(C)[n+](F)c(C)c1

InChI

1S/C8H11FN.BF4/c1-6-4-7(2)10(9)8(3)5-6;2-1(3,4)5/h4-5H,1-3H3;/q+1;-1

InChI key

RRNLYYDDEUOXBB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A source of electrophilic fluorine, used recently in the synthesis of fluoroazulenes.
Reactant for:
  • Pd(II)-catalyzed para-selective C-H arylation
  • Fluorination reactions
  • Aryl trifluoromethylation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bulletin of the Chemical Society of Japan, 69, 1645-1645 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service