Skip to Content
Merck
All Photos(1)

Key Documents

175064

Sigma-Aldrich

Nitrosyl tetrafluoroborate

95%

Synonym(s):

Nitrosonium tetrafluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NOBF4
CAS Number:
Molecular Weight:
116.81
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: oxidant

storage temp.

2-8°C

SMILES string

N#[O+].F[B-](F)(F)F

InChI

1S/BF4.NO/c2-1(3,4)5;1-2/q-1;+1

InChI key

KGCNVGDHOSFKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nitrosyl tetrafluoroborate is an efficient nitrosating and diazotizing agent. It reacts with alcohols and secondary amines to yield alkyl nitrites and nitrosamines, respectively. It reacts with primary amines to yield diazonium tetrafluoroborates. NOBF4 is also a mild oxidant and commonly used for single electron transfer oxidation.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Mohr et al.
FEBS letters, 348(3), 223-227 (1994-07-18)
Previous studies have suggested that glyceraldehyde-3-phosphate dehydrogenase (GAPDH) undergoes covalent modification of an active site thiol by a NO.-induced [32P]NAD(+)-dependent mechanism. However, the efficacy of GAPDH modification induced by various NO donors was found to be independent of spontaneous rates
Electronically regulated thermally and light-gated electron transfer from anions to naphthalenediimides.
Guha, Samit et al.
Journal of the American Chemical Society, 133(39), 15256-15259 (2011)
S Mohr et al.
The Journal of biological chemistry, 274(14), 9427-9430 (1999-03-27)
S-Nitrosylation of protein thiol groups by nitric oxide (NO) is a widely recognized protein modification. In this study we show that nitrosonium tetrafluoroborate (BF4NO), a NO+ donor, modified the thiol groups of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) by S-nitrosylation and caused enzyme
J Li et al.
Biochemical and biophysical research communications, 240(2), 419-424 (1997-12-06)
The caspases are a family of at least 10 human cysteine proteases that participate in cytokine maturation and in apoptotic signal transduction and execution mechanisms. Peptidic inhibitors of these enzymes are capable of blocking cytokine maturation and apoptosis, demonstrating their
C Würth et al.
Nanoscale, 9(12), 4283-4294 (2017-03-16)
The rational design of brighter upconversion nanoparticles (UCNPs) requires a better understanding of the radiationless deactivation pathways in these materials. Here, we demonstrate the potential of excitation power density (P)-dependent studies of upconversion (UC) luminescence intensities, slope factors, and absolute

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service