431052
Potassium metavanadate
98%
Synonym(s):
Potassium vanadate(V), Potassium vanadium trioxide
About This Item
Recommended Products
Quality Level
Assay
98%
form
powder
reaction suitability
reagent type: catalyst
core: vanadium
mp
520 °C (lit.)
density
2.84 g/mL at 25 °C (lit.)
SMILES string
[K+].[O-][V](=O)=O
InChI
1S/K.3O.V/q+1;;;-1;
InChI key
BQFYGYJPBUKISI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service