Skip to Content
Merck
All Photos(1)

Key Documents

425672

Sigma-Aldrich

Erbium(III) trifluoromethanesulfonate

98%

Synonym(s):

Erbium(III) triflate, Trifluoromethanesulfonic acid erbium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3)3Er
CAS Number:
Molecular Weight:
614.47
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

reaction suitability

core: erbium
reagent type: catalyst

SMILES string

[Er+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Er/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

GLQOFBCJADYRKR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

A water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service