Skip to Content
Merck
Erbium(III) trifluoromethanesulfonate 98%

Erbium(III) trifluoromethanesulfonate

Erbium(III) trifluoromethanesulfonate 98%
Synonym(s):
Erbium(III) triflate, Trifluoromethanesulfonic acid erbium(III) salt
Linear Formula:
(CF3SO3)3Er
CAS Number:
Molecular Weight:
614.47

Products

Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing