422894
2′,4′-Dimethylacetanilide
98%
Synonym(s):
2′,4′-Acetoxylidide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
98%
mp
129-132 °C (lit.)
functional group
amide
SMILES string
CC(=O)Nc1ccc(C)cc1C
InChI
1S/C10H13NO/c1-7-4-5-10(8(2)6-7)11-9(3)12/h4-6H,1-3H3,(H,11,12)
InChI key
PJEIIBXJUDOMAN-UHFFFAOYSA-N
General description
2′,4′-Dimethylacetanilide (N-(2,4-dimethylphenyl)acetamide) is an acetanilide derivative. Its carcinogenic effect has been studied on the liver in suckling mice. Its boiling point and melting point have been determined.
Application
2′,4′-Dimethylacetanilide (N-acetyl-2,4-xylidine) may be used as a starting material in the synthesis of 5-acetamido-2,4-dimethylaniline.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Carcinogenic effect of N-hydroxy-N-2-fluorenylacetamide, 2',4'-dimethylacetanilide, and 2',4'6'-trimethylacetanilide on liver in suckling mice.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 122(1), 111-114 (1966-05-01)
CRC Handbook of Chemistry and Physics, 3-218 (2015)
The Conversion of Liquid Rocket Fuels, 165, 125-133 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service