Skip to Content
Merck
All Photos(2)

Key Documents

187186

Sigma-Aldrich

Cinnamyl chloride

95%

Synonym(s):

(3-Chloropropenyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCH2Cl
CAS Number:
Molecular Weight:
152.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.584 (lit.)

bp

108 °C/12 mmHg (lit.)

mp

−19 °C (lit.)

density

1.096 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC\C=C\c1ccccc1

InChI

1S/C9H9Cl/c10-8-4-7-9-5-2-1-3-6-9/h1-7H,8H2/b7-4+

InChI key

IWTYTFSSTWXZFU-QPJJXVBHSA-N

General description

Cinnamyl chloride reacts regioselectively with aryl and alkenylgold(I) phosphanes in the presence of palladium catalyst in THF to afford the α-substitution product†.

Application

Cinnamyl chloride was used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Carlos Rueda et al.
Polymers, 12(6) (2020-06-26)
Stiff thermosensitive hydrogels (HG) were synthesized by self-crosslinking free radical polymerization of N,N-dimethylacrylamide (DMAA) and N-isopropylacrylamide (NIPAAm), adjusting the degree of swelling by carboxylate-containing sodium acrylate (NaAc) or a 2-oxazoline macromonomer (MM). The formation of hydrogels was possible due to
Wibke Lölsberg et al.
Organic letters, 14(23), 5996-5999 (2012-11-15)
A short and enantioselective total synthesis of helioporins C and E, which are bioactive marine diterpenes containing a serrulatane or amphilectane skeleton, was elaborated. The chirogenic step, i.e. a Cu(I)-catalyzed allylic alkylation of a cinnamyl chloride with methylmagnesium bromide, proceeded
Miguel Peña-López et al.
Organic & biomolecular chemistry, 10(8), 1686-1694 (2012-01-24)
Aryl and alkenylgold(I) phosphanes react regioselectively with allylic electrophiles such as cinnamyl and geranyl halides (bromide, chloride and acetates) under palladium catalysis in THF at 80 °C to afford the α-substitution product with moderate to high yields. When the reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service