Recommended Products
Quality Level
Assay
95%
form
liquid
refractive index
n20/D 1.5240 (lit.)
bp
218-220 °C (lit.)
density
1.273 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(cc1)[Si](Cl)(Cl)Cl
InChI
1S/C7H7Cl3Si/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3
InChI key
WOMUGKOOLXQCTQ-UHFFFAOYSA-N
Application
Self-assembled monolayers (SAMs) of p-tolyltrichlorosilane (TTCS) were formed on flexible polymer substrates. p-Tolyltrichlorosilane copolymerized with methylphenyldichlorosilane was grafted on silicon to introduce a structural defect.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
210.2 °F - closed cup
Flash Point(C)
99 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Effects of structural defects on hole drift mobility in aryl-substituted polysilanes.
Philosophical Magazine Part B, 79(10), 1631-1645 (1999)
Molecules (Basel, Switzerland), 25(13) (2020-07-02)
Novel polystyrene (PS)/polyhedral oligomeric silsequioxanes (POSSs) nanocomposites were designed and prepared by in situ polymerization, using, for the first time, three-cage POSS molecules. The synthesized compounds were first characterized by Fourier transform infrared spectroscopy (FTIR) and 1H NMR spectroscopy to
Fabrication of self-assembled monolayers (SAMs) and inorganic micropattern on flexible polymer substrate.
Langmuir, 20(8), 3278-3283 (2004)
Polymers, 12(11) (2020-11-25)
A comprehensive kinetics degradation study is carried out on novel multiple cages polyhedral oligomeric silsesquioxane (POSS)/polystyrene (PS) composites at 5% (w/w) of POSS to assess their thermal behavior with respect to the control PS and other similar POSS/PS systems studied
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service