Recommended Products
form
powder
Quality Level
mol wt
average Mn ~30,000
extent of labeling
12-15 wt. % Acetyl
36-40 wt. % Butyryl
refractive index
n20/D 1.475 (lit.)
density
1.25 g/mL at 25 °C (lit.)
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Masui. The Japanese journal of anesthesiology, 52(1), 26-31 (2003-03-14)
Dental injuries are the most common reason for complaints against anesthetists. The purpose of this study was to investigate the effect of teeth protector on dental injuries during general anesthesia. Incidence of dental injuries was evaluated retrospectively in 5,946 consecutive
Journal of pharmaceutical and biomedical analysis, 35(4), 779-788 (2004-06-15)
The purpose of this study was to qualitatively and quantitatively determine potential cellulose acetate butyrate (CAB) extractables in a way to meaningfully predict the in vivo exposure resulting from clinical administration. Extractions of CAB-381-20 were performed in several solvent systems
Journal of biomedical materials research. Part A, 101(1), 115-122 (2012-07-25)
Cellulose acetate butyrate nanofibers were prepared separately by two electrospinning techniques; a needleless electrospinning using a disc as spinneret and a rotary drum as collector and a conventional needle electrospinning using a rotary drum as collector. Compared to the needle-electrospun
AAPS PharmSciTech, 10(1), 147-157 (2009-02-10)
The purpose of this study was to formulate budesonide (BUD) compression-coated tablets for colonic specific delivery. Pectin and guar gum were used as enzyme-dependent polymers. For comparison purposes, both pH- and time-dependent polymers were also tried. In vitro release studies
Macromolecular bioscience, 5(9), 840-852 (2005-09-02)
Miscible blends of PHB and CAB were prepared by the solvent-casting method with various blend compositions, and their orientation behavior was investigated during uniaxial drawing. X-ray analysis revealed that the orientation of the crystallizable PHB component in the drawn PHB/CAB
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service