Skip to Content
Merck
All Photos(3)

Key Documents

247499

Sigma-Aldrich

Ethyl cellulose

viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder or granules

autoignition temp.

698 °F

concentration

48.0-49.5%

extent of labeling

48% ethoxyl

refractive index

n20/D 1.47 (lit.)

viscosity

100 cP, 5 % in toluene/ethanol 80:20(lit.)

transition temp

softening point 155 °C

density

1.14 g/mL at 25 °C (lit.)

SMILES string

O1[C@H](C(C(C(C1COCC)OCC)OCC)OCC)O[C@@H]2C(OC(C(C2OCC)OCC)OCC)COCC

InChI

1S/C28H54O11/c1-9-29-17-19-21(31-11-3)23(32-12-4)26(35-15-7)28(38-19)39-22-20(18-30-10-2)37-27(36-16-8)25(34-14-6)24(22)33-13-5/h19-28H,9-18H2,1-8H3/t19?,20?,21?,22-,23?,24?,25?,26?,27?,28+/m1/s1

InChI key

MINRZDZESGXKDM-OLIDUAMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl cellulose is a non-toxic, physiologicallyinert, and stable hydrophobic polymer. It is widely used in the preparation ofsustained release matrices for drug delivery systems.

Application

Ethyl cellulose can be used:
  • To prepare polymeric microcapsules of Alachlor and Metolachlor to study their controlled-release properties.
  • As a core matrix to synthesize biphasic drug release nanofibers.
  • To prepare wax-based solid dispersion pellets with excellent floating and sustained-release ability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Päivi Grönroos et al.
Sensors (Basel, Switzerland), 19(12) (2019-06-30)
Novel hot electron-emitting working electrodes and conventional counter electrodes were created by screen printing. Thus, low-cost disposable electrode chips for bioaffinity assays were produced to replace our older expensive electrode chips manufactured by manufacturing techniques of electronics from silicon or
Robert Morhard et al.
IEEE transactions on bio-medical engineering, 67(8), 2337-2348 (2019-12-17)
Ethanol ablation, the injection of ethanol to induce necrosis, was originally used to treat hepatocellular carcinoma, with survival rates comparable to surgery. However, efficacy is limited due to leakage into surrounding tissue. To reduce leakage, we previously reported incorporating ethyl
Anurag Verma et al.
Acta pharmaceutica (Zagreb, Croatia), 62(2), 237-250 (2012-07-04)
Chitosan has become a focus of major interest in recent years due to its excellent biocompatibility, biodegradability and non-toxicity. Although this material has already been extensively investigated in the design of different types of drug delivery systems, it is still
N Suwannateep et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 82(3), 485-490 (2012-09-08)
The phytochemical curcumin possesses antioxidant activity; however, it becomes unstable after being exposed to light or heat or loses activity during storage. This is especially important when curcumin is applied to the skin within a cosmetic or pharmaceutical formulation, since
Kifayat Ullah Shah et al.
TheScientificWorldJournal, 2012, 842348-842348 (2012-06-01)
The design and fabrication of sustained/controlled release dosage forms, employing new excipients capable of extending/controlling the release of drugs from the dosage forms over prolonged periods, has worked well in achieving optimally enhanced therapeutic levels of the drugs. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service