All Photos(1)
About This Item
Linear Formula:
Cl(CH2)8OH
CAS Number:
Molecular Weight:
164.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.458 (lit.)
bp
129-130 °C/11 mmHg (lit.)
density
0.976 g/mL at 25 °C (lit.)
functional group
chloro
hydroxyl
SMILES string
OCCCCCCCCCl
InChI
1S/C8H17ClO/c9-7-5-3-1-2-4-6-8-10/h10H,1-8H2
InChI key
YDFAJMDFCCJZSI-UHFFFAOYSA-N
Related Categories
General description
8-Chloro-1-octanol is a ω-chloro-1-alkanol.
Application
8-Chloro-1-octanol may be employed for the following studies:
- Biosynthesis of bacteriochlorophyll c derivatives, via esterification.
- Preparation of new macroporous triisobutylphosphine sulphide functionalized polymers.
- Synthesis of series of new mesogenic azomethine diols.
- Synthesis of bis(4-hydroxyoctoxyphenyl)sulfone (HOS).
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
New macroporous polymers for the selective adsorption of gold (III) and palladium (II). I. The synthesis, characterization, and effect of spacers on metal adsorption.
Sanchez JM, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 38(2), 269-278 (2000)
Issam Ahmed Mohammed et al.
Molecules (Basel, Switzerland), 15(5), 3260-3269 (2010-07-27)
A series of new mesogenic azomethine diols were successfully synthesized by condensation reactions between various chloroalkanols and N,N'-bis(4-hydroxy)-benzylidene-o-toluidine (1). The structures of these compounds were confirmed by CHN, FT-IR, (1)H-NMR, and (13)C-NMR spectrophotometer. Their thermotropic liquid crystalline behavior was studied
Synthesis and Properties of a Ferrocene-based Metallomesogenic Polymer Containing Bis (4-hydroxyoctoxyphenyl) sulfone.
Amer WA, et al.
Journal of Inorganic and Organometallic Polymers and Materials, 22(6), 1229-1239 (2012)
Yoshitaka Saga et al.
Journal of bioscience and bioengineering, 118(1), 82-87 (2014-02-06)
The green sulfur photosynthetic bacterium Chlorobaculum tepidum newly produced BChl c derivatives possessing a chlorine or bromine atom at the terminus of the esterifying chain in the 17-propionate residue by cultivation with exogenous ω-halo-1-alkanols. The relative ratios of BChl c
Zachary K Zander et al.
Biomaterials, 178, 339-350 (2018-05-23)
The use of catheters is ubiquitous in medicine and the incidence of infection remains unacceptably high despite numerous advances in functional surfaces and drug elution. Herein we report the use of a thermoplastic polyurethane containing an allyl ether side-chain functionality
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service