Skip to Content
Merck
All Photos(1)

Key Documents

278823

Sigma-Aldrich

4-Chloro-1-butanol

technical grade, ~85%

Synonym(s):

Tetramethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)4Cl
CAS Number:
Molecular Weight:
108.57
Beilstein:
1731408
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

~85%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

84-85 °C/16 mmHg (lit.)

density

1.088 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

OCCCCCl

InChI

1S/C4H9ClO/c5-3-1-2-4-6/h6H,1-4H2

InChI key

HXHGULXINZUGJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-1-butanol, an alkylating agent, is one of the genotoxic impurity in active pharmaceutical ingredients. Reaction of 4-chloro-1-butanol and Reichardt′s dye has been monitored spectrophotometrically for the determination of genotoxic impurities in pharmaceuticals.

Application

4-Chloro-1-butanol was employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

Other Notes

Contains tetrahydrofuran and HCl

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Koki Harigaya et al.
Chemical & pharmaceutical bulletin, 62(4), 395-398 (2014-04-04)
The analysis of genotoxic impurities (GTIs) in active pharmaceutical ingredients (APIs) is a challenging task. The target detection limit (DL) in an API is typically around 1 ppm (1 µg/g API). Therefore, a sensitive and selective analytical method is required for their
Damion K Corrigan et al.
The Journal of pharmacy and pharmacology, 61(4), 533-537 (2009-03-21)
Alkylating agents are potentially genotoxic impurities that may be present in drug products. These impurities occur in pharmaceuticals as by-products from the synthetic steps involved in drug production, as impurities in starting materials or from in-situ reactions that take place

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service