Merck
  • Production of Soda Lime Glass Having Antibacterial Property for Industrial Applications.
Production of Soda Lime Glass Having Antibacterial Property for Industrial Applications.
Materials (Basel, Switzerland) (2020-11-01)
Barış Demirel, Melek Erol Taygun
ABSTRACT
This study was aimed to produce and characterize the first commercial glass materials with enhanced antibacterial property using conventional melting method. For this purpose, typical container glass composition that contains some specific metal ions, such as silver, strontium, and copper, was used to obtain antibacterial glass samples using classical melting method. After the melting process, antibacterial tests and migration tests were applied to the glasses, and it was found that the glass doped with 2% Ag2O was the best composition. X-rays diffractometer (XRD), thermal expansion coefficient, density, refractive index, hardness, and elastic module results showed that the glass doped with 2% Ag2O was a suitable material as a container glass. High Temperature Melting Observation System studies were performed on the produced antibacterial glass composition, and it was found that the antibacterial glass can be produced in soda lime glass furnaces without changing any furnace design and production parameters. As a result of the characterization studies, it was concluded that the produced container glass doped with silver can be a good candidate for food and pharmaceutical products where bacterial growth is absolutely undesirable.
Sigma-Aldrich
Strontium oxide, 99.9% trace metals basis
Pricing and availability is not currently available.