Skip to Content
Merck
All Photos(1)

Key Documents

411760

Sigma-Aldrich

Furfuryl methacrylate

97%, contains 200 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Methacrylic Acid Furfuryl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid

contains

200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.482 (lit.)

bp

80-82 °C/5 mmHg (lit.)

density

1.078 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCc1ccco1

InChI

1S/C9H10O3/c1-7(2)9(10)12-6-8-4-3-5-11-8/h3-5H,1,6H2,2H3

InChI key

DWXAVNJYFLGAEF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunanda Sain et al.
Polymers, 12(11) (2020-10-30)
The fabrication of smart biocomposites from sustainable resources that could replace today's petroleum-derived polymer materials is a growing field of research. Here, we report preparation of novel biocomposites using nanocellulose networks extracted from food residue (onion skin) and a vegetable
Sunanda Sain et al.
Polymers, 12(2) (2020-01-26)
This work focuses on the development of cross-linked polymer from a highly unsaturated vegetable oil, tung oil (TO) and a bio-based acrylate, furfuryl methacrylate (FMA). The presence of a high degree of unsaturated carbon-carbon bonding in TO makes it a
A Amalin Kavitha et al.
ACS applied materials & interfaces, 1(7), 1427-1436 (2010-04-02)
This investigation reports the effective use of the Diels-Alder (DA) reaction, a "click reaction" in the preparation of thermally amendable and self-healing polymeric materials having reactive furfuryl functionality. In this case, the DA and retro-DA (rDA) reactions were carried out
Chuanwei Lu et al.
Carbohydrate polymers, 242, 116404-116404 (2020-06-23)
Well-defined sustainable and recyclable thermoset elastomers derived from cellulose, fatty acid and furfural were successfully prepared via photoinduced metal-free ATRP and Diels-Alder (DA) reaction. Firstly, metal free ATRP was applied to prepare a range of thermoplastic cellulose graft copolymers with
D Zaldívar et al.
Biomaterials, 14(14), 1073-1079 (1993-11-01)
Biocompatible copolymers of N-vinylpyrrolidone (P) and furfuryl methacrylate (F) were prepared by free radical polymerization in N,N-dimethylformamide solution at 50 degrees C, using 2,2'-azobisisobutyronitrile as initiator, at low and high conversion. The microstructure of copolymers prepared at low conversion was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service