Skip to Content
Merck
All Photos(2)

Key Documents

773263

Sigma-Aldrich

N-(2-Hydroxyethyl)maleimide

97%

Synonym(s):

1-(2-Hydroxyethyl)-pyrrole-2,5-dione, 1-(2-Hydroxyethyl)maleimide, N-(Ethanol)maleimide, HEMI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO3
CAS Number:
Molecular Weight:
141.12
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: cross-linking reagent

mp

66-67 °C (lit.)
67-74 °C

functional group

hydroxyl
maleimide

storage temp.

2-8°C

SMILES string

O=C(N1CCO)C=CC1=O

InChI

1S/C6H7NO3/c8-4-3-7-5(9)1-2-6(7)10/h1-2,8H,3-4H2

InChI key

AXTADRUCVAUCRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-(2-Hydroxyethyl)maleimide can be used:
  • In the synthesis of (EFA)-based multifunctional oligoester resins with maleimides as end groups by reacting with 9,10-epoxy-18-hydroxyoctadecanoic acid (EFA) and dimethyl adipate catalyzed by Candida antarctica lipase B (CalB).
  • As an initiator in ring-opening polymerization of δ-valerolactone catalyzed by trifluoromethanesulfonimide.
  • In one-pot synthesis of bio-based polyurethane-imides by reacting with castor oil, eleostearic acid diethanol amide and isophorone diisocyanate.

N-(2-Hydroxyethyl)maleimide may be used to synthesize [2,2-bis(maleimidoethoxy)-
propane (BMEP)], a protein cross-linking reagent with potential application in constructing immunotoxins. It may also be used in preparing thermoresponsive self-healing polyurethanes with the shape-memory property.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot synthesis of cyclopentadienyl endcapped poly (2-ethyl-2-oxazoline) and subsequent ambient temperature Diels?Alder conjugations.
Glassner M, et al.
Chemical Communications (Cambridge, England), 47(38), 10620-10622 (2011)
Enzymatic catalysis as a versatile tool for the synthesis of multifunctional, bio-based oligoester resins
Semlitsch S, et al.
Green Chemistry, 18(7), 1923-1929 (2016)
One-pot synthesis of polyurethane-imides with tailored performance from castor and tung oil
Feng Y, et al.
Progress in Organic Coatings, 132(17), 62-69 (2019)
New protein crosslinking reagents that are cleaved by mild acid.
Srinivasachar K & Neville Jr DM.
Biochemistry, 28(6), 2501-2509 (1989)
Self?Healing Polyurethanes with Shape Recovery.
Heo Y & Sodano HA.
Advances in Functional Materials, 24(33), 5261-5268 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service