411140
(S)-(+)-α-(Trifluoromethyl)benzyl alcohol
99%
Synonym(s):
(+)-Phenyl(trifluoromethyl)carbinol, (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol
About This Item
Recommended Products
Assay
99%
form
liquid
optical activity
[α]20/D +31.3±0.5°, neat
optical purity
ee: 97% (GLC)
refractive index
n20/D 1.462 (lit.)
bp
73-76 °C/9 mmHg (lit.)
density
1.3 g/mL at 25 °C (lit.)
functional group
fluoro
hydroxyl
phenyl
SMILES string
O[C@@H](c1ccccc1)C(F)(F)F
InChI
1S/C8H7F3O/c9-8(10,11)7(12)6-4-2-1-3-5-6/h1-5,7,12H/t7-/m0/s1
InChI key
VNOMEAQPOMDWSR-ZETCQYMHSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
183.2 °F - closed cup
Flash Point(C)
84 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service