Skip to Content
Merck
All Photos(1)

Key Documents

405965

Sigma-Aldrich

2,2′-Methylenebis[(4S)-4-tert-butyl-2-oxazoline]

99%

Synonym(s):

(S,S)-2,2′-Methylenebis(4-tert-butyl-2-oxazoline)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26N2O2
CAS Number:
Molecular Weight:
266.38
Beilstein:
4190673
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −118°, c = 0.5 in chloroform

mp

51-53 °C (lit.)

functional group

ether

SMILES string

CC(C)(C)[C@H]1COC(CC2=N[C@H](CO2)C(C)(C)C)=N1

InChI

1S/C15H26N2O2/c1-14(2,3)10-8-18-12(16-10)7-13-17-11(9-19-13)15(4,5)6/h10-11H,7-9H2,1-6H3/t10-,11-/m1/s1

InChI key

WCCCBUXURHZPQL-GHMZBOCLSA-N

Application

2,2′-Methylenebis[(4S)-4-tert-butyl-2-oxazoline] (BOX ligand) can be used as a C2 symmetric ligand for enantioselective asymmetric catalysis. It easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.
C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneous bisoxazoline/copper complex: A green catalyst for the enantioselective reaction of nitromethane with substituted benzaldehydes
Maggi R, et al.
European Journal of Organic Chemistry, 2011(28), 5551-5554 (2011)
Selective oxidation of alkenes and alkynes catalyzed by copper complexes
Alvarez LX, et al.
Applied Catalysis A: General, 325(2), 303-308 (2007)
C 2-Symmetric bis (oxazolinato) lanthanide catalysts for enantioselective intramolecular hydroamination/cyclization
Hong S, et al.
Journal of the American Chemical Society, 125(48), 14768-14783 (2003)
Gant, T.G. Meyers, A.I.
Tetrahedron, 50, 2297-2297 (1994)
Bolm, C.
Angewandte Chemie (International Edition in English), 30, 542-542 (1991)

Articles

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service