Skip to Content
Merck
All Photos(1)

Key Documents

404888

Sigma-Aldrich

1-Methyl-2-phenylindole

99%

Synonym(s):

NSC 63793

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N
CAS Number:
Molecular Weight:
207.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

mp

98-100 °C (lit.)

SMILES string

Cn1c(cc2ccccc12)-c3ccccc3

InChI

1S/C15H13N/c1-16-14-10-6-5-9-13(14)11-15(16)12-7-3-2-4-8-12/h2-11H,1H3

InChI key

SFWZZSXCWQTORH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 1-methyl-2-phenylindole with nitrogen dioxide or with nitrous acid (NaNO2-CH3COOH) in benzene has been studied. 1-Methyl-2-phenylindole is reported to react with malondialdehyde (MDA) and 4-hydroxyalkenals to afford a stable chromophore with intense maximal absorbance at 586nm.

Application

1-Methyl-2-phenylindole may be used in the following studies:
  • The estimation of lipid peroxidation in third instar larvae of transgenic Drosophila melanogaster (hsp70-lacZ)Bg.
  • As chromogenic agent for the determination of estimation of malondialdehyde (MDA) production.
  • Colorimetric assay of lipid peroxidation.
  • Synthesis of 1-methyl-2-phenyl-3-(1,3,4-thiadiazol-2-yldiazenyl)-1H-indole.
  • Synthesis of 3-(5-ethyl-1,3,4-thiadiazol-2-yldiazenyl)-1-methyl-2-phenyl-1H-indole.
Reactant for preparation of:
  • Cyano indoles
  • Difluorohydroxy indoles

Reactant for:
  • Carboxylation of indoles
  • Allylation of indoles
  • Formylation of indoles
  • Nitrosylation of indoles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Gérard-Monnier et al.
Chemical research in toxicology, 11(10), 1176-1183 (1998-10-20)
Under acidic and mild-temperature conditions, 1-methyl-2-phenylindole was found to react with malondialdehyde (MDA) and 4-hydroxyalkenals to yield a stable chromophore with intense maximal absorbance at 586 nm. The use of methanesulfonic acid results in optimal yields of chromophore produced from
Bread Leandro Gomes Cruz et al.
BMC cancer, 16, 58-58 (2016-02-06)
The occurrence of cancer during pregnancy merges two complex, poorly understood metabolic and hormonal conditions. This association can exacerbate the conditions of both the mother and the foetus. The branched-chain amino acid leucine enhances cellular activity, particularly by increasing protein
Marcin Magierowski et al.
Journal of gastroenterology, 53(1), 52-63 (2017-02-27)
Aspirin exerts side effects within the gastrointestinal tract. Hydrogen sulfide (H Wistar rats with or without capsaicin-induced denervation of sensory neurons were pretreated with vehicle, CORM-2 (5 mg/kg intragastrically), or NaHS (5 mg/kg intragastrically) with or without capsazepine (5 mg/kg intragastrically) or N
Abraham Said Arellano Buendía et al.
International journal of molecular sciences, 19(10) (2018-10-14)
Diabetic nephropathy (DN) is presently the primary cause of chronic kidney disease and end-stage renal disease (ESRD). It has been suggested that inflammation and oxidative stress, in addition to or in concert with the metabolic changes, plays an important role
María Hernández-Trejo et al.
BMC immunology, 18(1), 3-3 (2017-01-08)
Oxidative damage present in obese/overweight mothers may lead to further oxidative stress conditions or inflammation in maternal and cord blood samples. Thirty-four pregnant women/newborn pairs were included in this study to assess the presence of oxidative stress biomarkers and their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service