Skip to Content
Merck
All Photos(1)

Key Documents

402168

Sigma-Aldrich

6-Bromo-3-formylchromone

99%

Synonym(s):

6-Bromo-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5BrO3
CAS Number:
Molecular Weight:
253.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

190-193 °C (lit.)

functional group

aldehyde
bromo
ketone

SMILES string

[H]C(=O)C1=COc2ccc(Br)cc2C1=O

InChI

1S/C10H5BrO3/c11-7-1-2-9-8(3-7)10(13)6(4-12)5-14-9/h1-5H

InChI key

PCEZXSJBHMOQFT-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

General description

6-Bromo-3-formylchromone (6-Bromo-4-oxo-4H-1-benzopyran-3-carboxaldehyde) is a 3-formylchromone derivative. In vivo cytotoxic activity of 6-bromo-3-formylchromone against normal and tumor cells has been tested.

Application

6-Bromo-3-formylchromone is the suitable reagent used in a study to investigate the multidrug resistance reversal by some 3- formylchromones in human colon cancer and mouse lymphoma cells transfected with the human MDR1 gene. It may be used in the preparation of chromone containing sulfonamides.
6-Bromo-3-formylchromone may be used in the preparation of 6′-bromopyranothiazine-4,7-diones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A study of the reactions of 2-aryl-4-hydroxy-6H-1,3-thiazin-6-ones with chromone-3-carboxaldehydes.
Shutov RV, et al.
Tetrahedron Letters, 52(2), 266-269 (2011)
Farukh Arjmand et al.
Bioorganic chemistry, 104, 104327-104327 (2020-11-05)
Copper-based antitumor drug entities 1-3 derived from substituted (F-, Br-, -CH3) 3-formylchromone pharmacophore were synthesized and thoroughly characterized by spectroscopic and single X-ray crystallographic studies. These complexes show structural novelty due to presence of the X-bonds in chromone scaffold which
Mariya al-Rashida et al.
Bioorganic & medicinal chemistry, 19(11), 3367-3371 (2011-05-11)
Series of chromone containing sulfonamides were prepared by the reaction of (un)substituted 3-formylchromones with 3-aminobenzenesulfonamide and 4-aminobenzenesulfonamide. Bovine carbonic anhydrase (bCA) inhibitory activity of these newly synthesized compounds was determined. All compounds were active and possessed excellent bCA inhibitory activities
Masami Kawase et al.
In vivo (Athens, Greece), 21(5), 829-834 (2007-11-21)
Several 3-formylchromone derivatives were examined for their tumor cell-cytotoxic, anti-Helicobacter pylori, urease inhibitory and anti-HIV activity. Comparing their relative cytotoxicity against four human tumor cell lines and three normal human cells, tumor cell-specific cytotoxicity was detected in some 3-formylchromone derivatives.
Zoltán Baráth et al.
In vivo (Athens, Greece), 20(5), 645-649 (2006-11-10)
Several new 3-formylchromone derivatives proved to be modifiers of multidrug resistance in mouse lymphoma cells and in human Colo320 colon cancer cells. There is apparently a structure-activity relationship between the antiproliferative multidrug resistance-reversing effect and the chemical structure of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service