Skip to Content
Merck
All Photos(3)

Key Documents

400904

Sigma-Aldrich

Lithium molybdate

greener alternative

99.9% trace metals basis

Synonym(s):

Dilithium dioxido(dioxo)molybdenum, Lithium Molybdate(VI), Lithium Molybdenum Oxide, Lithium metamolybdate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2MoO4
CAS Number:
Molecular Weight:
173.82
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
powder or crystals

Quality Level

Assay

99.9% trace metals basis

form

powder or crystals

reaction suitability

reagent type: catalyst
core: molybdenum

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1500.0 ppm Trace Metal Analysis

density

2.66 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].[Li+].[O-][Mo]([O-])(=O)=O

InChI

1S/2Li.Mo.4O/q2*+1;;;;2*-1

InChI key

NMHMDUCCVHOJQI-UHFFFAOYSA-N

General description

Lithium molybdate is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium molybdate can be used as an electrode material, a corrosion inhibitor, and a refrigerant. It has a three-dimensional framework that is useful in the fabrication of lithium-ion batteries. It facilitates the improvement of rate capability and cyclic stability of the electrochemical system.
Used in comparative property studies of battery systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization and electrochemical properties of Li2MoO4 modified Li4Ti5O12/C anode material for lithium-ion batteries
Liu J, et al.
Electrochimica Acta, 170, 202-209 (2015)
Electrical transport properties of semiconducting lithium molybdate glass nanocomposites
Bhattacharya S and Ghosh A
J. Chem. Phys. , 127(19), 194709-194709 (2007)
Dielectric properties of lithium molybdate ceramic fabricated at room temperature
Kahari H, et al.
Journal of the American Ceramic Society. American Ceramic Society, 97(11), 3378-3379 (2014)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service