Skip to Content
Merck
All Photos(1)

Key Documents

8.00408

Sigma-Aldrich

Maleic anhydride

for synthesis

Synonym(s):

Maleic anhydride, 2,5-Furanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2O3
CAS Number:
Molecular Weight:
98.06
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-571-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 44 °C)

Quality Level

form

solid

autoignition temp.

475 °C (DIN 51794)

potency

1090 mg/kg LD50, oral (Rat)
2620 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-7.1 % (v/v)

pH

0.8 (20 °C, 550 g/L in H2O, Hydrolysis)

bp

202 °C/1013 hPa

mp

51-53 °C

transition temp

flash point 103 °C

density

1.32 g/cm3 at 55 °C

bulk density

700‑800 kg/m3

storage temp.

2-30°C

SMILES string

O1C(=O)C=CC1=O

InChI

1S/C4H2O3/c5-3-1-2-4(6)7-3/h1-2H

InChI key

FPYJFEHAWHCUMM-UHFFFAOYSA-N

Application

  • Chemical modification of polypropylene by maleic anhydride: This study explored the application of maleic anhydride (MAH) for modifying the properties of polypropylene through melt grafting. The findings reveal how variations in the concentration of MAH influence the polymer′s properties (Ebadi-Dehaghani et al., 2014).
  • Renewable production of phthalic anhydride: A study on the synthesis of phthalic anhydride from biomass-derived furan and maleic anhydride, highlighting an eco-friendly pathway for producing a widely used industrial chemical (Mahmoud et al., 2014).
  • Enhancement of biopolymer properties: Research on using maleic anhydride as a compatibilizer in natural fiber-reinforced poly(lactic acid) composites, which demonstrates the improvements in mechanical and thermal properties of the composites (Yu et al., 2014).
  • Application in electrical property enhancement: This article reports on the application of maleic anhydride grafted polypropylene which significantly enhances the material′s electrical properties, potentially applicable in various advanced electronics and engineering fields (Zhou et al., 2016).

Analysis Note

Assay (morpholine method): ≥ 99,0 %(m)
Melting range (lower value): ≥ 51 °C
Melting range (upper value): ≤ 54 °C
Identity (IR): conforms

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1A - STOT RE 1 Inhalation

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

217.4 °F

Flash Point(C)

103 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service