Skip to Content
Merck
All Photos(1)

Key Documents

396788

Sigma-Aldrich

Ethyl formimidate hydrochloride

Synonym(s):

Ethyl methanimidate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC(=NH)OC2H5·HCl
CAS Number:
Molecular Weight:
109.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

75 °C (dec.) (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

Cl.CCOC=N

InChI

1S/C3H7NO.ClH/c1-2-5-3-4;/h3-4H,2H2,1H3;1H

InChI key

JPUTTYRVDANTBN-UHFFFAOYSA-N

General description

Ethyl formimidate hydrochloride is a nitrogen containing organic building block. On heating it undergoes degradation to afford formamidine hydrochloride, ethyl formate and ethyl chloride.

Application

Ethyl formimidate hydrochloride may be used in the preparation of the following:
  • 5-aminoimidazole-4-carboxylic acid α- and β-ribotides
  • s-triazines
  • 2,3:5,6-di-O-isopropylidene-α- and -β-5-amino-4-ethoxycarbonyl or -carbamoyl-imidazole D-mannofuranosides
  • 5-amino-1-(2-pyridyl)imidazole
Reactant involved in the synthesis of biologically active molecules including:
  • Bredinin via amination of an acyclic precursor
  • Amidine conjugates of the ornithine moiety of an antifungal

Reactant involved in:
  • Intermolecular cyclization
  • Mo-catalyzed asymmetric ring-closing metathesis for synthesis of cyclid amides and amines
  • Synthesis of peptidic 1-cyanopyrrolidines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purines, pyrimidines, and imidazoles. XL. A new synthesis of a D-ribofuranosylamine derivative and its use in the synthesis of pyrimidine and imidazole nucleosides.
N J Cusack et al.
Journal of the Chemical Society. Perkin transactions 1, 16, 1720-1731 (1973-01-01)
Purines, pyrimidines, and imidazoles. Part XLI. Glycofuranosylamines derived from D-xylose, D-glucose, D-mannose, and L-rhamnose and their use in the synthesis of pyrimidine and imidazole nucleosides.
Cusack NJ, et al.
Journal of the Chemical Society. Perkin Transactions 1, 73-81 (1974)
Synthesis of the S-Triazine System. Iii. 1 Trimerization of Imidates.
Schaefer FC and Peters GA.
The Journal of Organic Chemistry, 26(8), 2771-2784 (1961)
Rhodd EH and von Richter V.
Chemistry of Organic Compounds: Pt. B. Aliphatic compounds, 551-551 (1951)
Purines, pyrimidines, and imidazoles part 54. Interconversion of some intermediates in the de novo biosynthesis of purine nucleotides.
Cusack NJ, et al.
Journal of the Chemical Society. Perkin Transactions 1, 2316-2321 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service