Skip to Content
Merck
All Photos(1)

Key Documents

391107

Sigma-Aldrich

trans-1-Bromo-1-propene

contains copper as stabilizer, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHBr
CAS Number:
Molecular Weight:
120.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

contains

copper as stabilizer

refractive index

n20/D 1.453 (lit.)

bp

64-65 °C (lit.)

density

1.408 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

storage temp.

2-8°C

SMILES string

C\C=C\Br

InChI

1S/C3H5Br/c1-2-3-4/h2-3H,1H3/b3-2+

InChI key

NNQDMQVWOWCVEM-NSCUHMNNSA-N

General description

trans-1-Bromo-1-propene is an alkenyl halide. Product contains copper as stabilizer. Its synthesis from 1,2-dibromopropane has been reported by various researchers. Its IR spectra has been investigated.

Application

trans -1-Bromo-1-propene is suitable for the synthesis of (E)-hex-4-en-2-yn-1-ol. trans -1-Bromo-1-propene ((E)-1-bromo-1-propene) may be used in the synthesis of the following:
  • (E)-1,3-dimethyl-3-(prop-1-enyl)indolin-2-one
  • 3-alkenyl-Δ3-cephems
  • (E)-1-(benzylthio)-1-propene
trans-1-Bromo-1-propene may be used as starting reagent in the synthesis of (+)-trans-isoalliin and (-)-trans-isoalliin.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

5.0 °F - closed cup

Flash Point(C)

-15 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure Analysis of Poly (propylene-β-d oxide) by Proton Nuclear Magnetic Resonance Spectroscopy.
Oguni N, et al.
Macromolecules, 6(3), 459-465 (1973)
Lasanthi Jayathilaka et al.
Journal of biomolecular techniques : JBT, 25(3), 67-76 (2014-09-05)
Naturally occurring (+)-trans-isoalliin, (R(C)R(S))-(+)-trans-S-1-propenyl-L-cysteine sulfoxide, is a major cysteine sulfoxide in onion. The importance of producing it synthetically to support further research is very well recognized. The (+)-trans-isoalliin is prepared by chemical synthesis and reversed-phase (RP)-HPLC. First, S-2-propenyl-L-cysteine (deoxyalliin) is
Substituted Dienols from Palladium Catalyzed Coupling of Hydroaluminated Enynols with Aryl Iodides.
Crook KE, et al.
Letters in Organic Chemistry, 5(3), 158-164 (2008)
H Tanaka et al.
The Journal of organic chemistry, 66(2), 570-577 (2001-06-30)
Synthesis of 3-alkenyl-delta 3-cephems was performed successfully by cross-coupling 3-(trifluoromethylsulfonyloxy or chloro)-delta 3-cephem with alkenyl halides, e.g., vinyl bromide, trans-1-bromo-1-propene, and trans-beta-bromostyrene in an Al/cat.PbBr2/cat.NiBr2(bpy)/NMP (or DMF) system. Reduction of 3-(trifluoromethylsulfonyloxy)-delta 3-cephem into norcephalosporin was also achieved by a similar
Alexander M Taylor et al.
Journal of the American Chemical Society, 131(29), 9900-9901 (2009-07-08)
The enantioselective alpha-arylation and alpha-vinylation of oxindoles catalyzed by Pd and a biarylmonophosphine ligand with both axial and phosphorus-based chirogenicity is reported. The resultant quaternary carbon stereocenters are formed in high enantiomeric excess, and the conditions tolerate a range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service