Recommended Products
Assay
98%
mp
295-297 °C (lit.)
functional group
bromo
SMILES string
BrCc1c(CBr)c(CBr)c(CBr)c(CBr)c1CBr
InChI
1S/C12H12Br6/c13-1-7-8(2-14)10(4-16)12(6-18)11(5-17)9(7)3-15/h1-6H2
InChI key
XJOUCILNLRXRTF-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service