Skip to Content
Merck
All Photos(4)

Key Documents

125911

Sigma-Aldrich

α,α′-Dibromo-m-xylene

greener alternative

97%

Synonym(s):

1,3-Bis(bromomethyl)benzene, m-Xylylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH2Br)2
CAS Number:
Molecular Weight:
263.96
Beilstein:
971085
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

greener alternative product score

old score: 10
new score: 6
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Atom Economy
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

135-140 °C/20 mmHg (lit.)

mp

74-77 °C (lit.)

functional group

bromo

greener alternative category

SMILES string

BrCc1cccc(CBr)c1

InChI

1S/C8H8Br2/c9-5-7-2-1-3-8(4-7)6-10/h1-4H,5-6H2

InChI key

OXHOPZLBSSTTBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α,α′-Dibromo-m-xylene can cross-link and cyclize peptides containing two cysteine residues.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy” and “Safer Solvents and Auxiliaries”. Click here to view its DOZN scorecard.

Application

α,α′-Dibromo-m-xylene has been used in preparation of four novel dinuclear half-titanocenes with meta- and orthoxylene bridges as new metallocenes. It was used in the synthesis of dissymmetrical bis-macrocyclic salt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian P Seebeck et al.
Journal of the American Chemical Society, 128(22), 7150-7151 (2006-06-01)
Dehydroalanine is a nonproteinogenic amino acid, but it is a component of a wide variety of natural products with therapeutic activities. Indeed, this alpha,beta-unsaturated residue is a highly versatile building block due to its rigidifying effect on peptide backbones and
Bis-aminals: efficient tools for bis-macrocycle synthesis.
Le Baccon M, et al.
New. J. Chem., 25(9), 1168-1174 (2001)
Preparation of new dinuclear half-titanocene complexes with ortho- and meta-xylene linkages and investigation of styrene polymerization.
Linh NTB, et al.
Journal of Organometallic Chemistry, 694(21), 3438-3443 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service