Skip to Content
Merck
All Photos(1)

Key Documents

386588

Sigma-Aldrich

Pinacolyl methylphosphonate

97%

Synonym(s):

1,2,2-Trimethylpropyl methylphosphonate, 3,3-Dimethylbutan-2-yloxy(methyl)phosphinic acid, Mono(1,2,2-trimethylpropyl) methylphosphonate, O-Pinacolyl hydrogen methylphosphonate, O-Pinacolyl methylphosphonate, PMPA, Pinacolyl hydrogen methylphosphonate, [(3,3-Dimethylbutan-2-yl)oxy](methyl)phosphinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(O)(OH)OCH(CH3)C(CH3)3
CAS Number:
Molecular Weight:
180.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

96-106 °C/8 mmHg (lit.)

density

1.032 g/mL at 25 °C (lit.)

SMILES string

CC(OP(C)(O)=O)C(C)(C)C

InChI

1S/C7H17O3P/c1-6(7(2,3)4)10-11(5,8)9/h6H,1-5H3,(H,8,9)

InChI key

BLALDUPQYCGKAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Looking down the drug pipeline.
J Cadman
GMHC treatment issues : the Gay Men's Health Crisis newsletter of experimental AIDS therapies, 12(3), 5-9 (2001-05-22)
Sung-Up Choi et al.
Journal of pharmaceutical sciences, 97(2), 931-943 (2007-06-05)
We have investigated the ability of lipid-indinavir particles composed of 3-to-1 lipid-drug molar ratio to encapsulate an aqueous marker calcein and anti-HIV drug (3)H-phosphonylmethoxypropyl-adenine (PMPA). Even at a high density of indinavir associated to lipid-indinavir nanoparticles, they form an enclosed
Manisha Sathe et al.
The Analyst, 137(2), 406-413 (2011-11-19)
Immunoassay detection of O-pinacolyl methylphosphonic acid (PMPA) employing direct coating of N-2-aminoethyl-O-pinacolyl methylphosphonate (hapten B) on microtiter plates is reported. Coating was achieved by covalently linking hapten B to a glutaraldehyde (GA) polymer network directly bound to the polystyrene (PS)
B Rosenwirth et al.
Journal of medical primatology, 28(4-5), 195-205 (1999-12-11)
Combination chemotherapy using potent anti-retroviral agents has led to significant advances in the clinical management of human immunodeficiency virus (HIV) disease. However, the emergence of multiple drug-resistant mutants, the high need for compliance to adhere to demanding drug-dosing schemes, and
Harri Koskela et al.
Analytical chemistry, 90(14), 8495-8500 (2018-06-12)
The NMR-observable nuclei of the acidic and basic compounds experience pH dependence in chemical shift. This phenomenon can be exploited in NMR titrations to determine p Ka values of compounds, or in pH measurement of solutions using dedicated pH reference

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service