Skip to Content
Merck
All Photos(2)

Key Documents

380237

Sigma-Aldrich

Tebbe reagent solution

0.5 M in toluene

Synonym(s):

Bis(cyclopentadienyl)-μ-chloro­(dimethylaluminum)-μ-methylenetitanium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18AlClTi
CAS Number:
Molecular Weight:
284.58
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: C-C Bond Formation

concentration

0.5 M in toluene

density

0.927 g/mL at 25 °C

SMILES string

[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.C[Al](C)C[Ti]Cl

InChI

1S/2C5H5.2CH3.CH2.Al.ClH.Ti/c2*1-2-4-5-3-1;;;;;;/h2*1-5H;2*1H3;1H2;;1H;/q;;;;;;;+1/p-1

InChI key

QEJAQNUJXFLWSP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tebbe reagent is a Lewis acid stabilized organometallic compound, which is used to transform carbonyl groups into β-substituted methylenes. It also reacts with a wide range of carbonyl compounds, including esters, amides, and lactones to yield corresponding olefins.

Application

Tebbe reagent can be used:     
  • For the conversion of carbonyl groups of chlorophyll derivatives into the corresponding exo-methylene (or vinylidene) groups.
  • In the synthesis of β-C-glycosides from 3-OH glycol esters.
  • As a versatile methylenation reagent for the conversion of ketones and aldehydes to olefins. It offers facile reaction with hindered ketones and allows the conversion of esters to vinyl ethers.
  • To olefinate aldehydes.
  • To methylenate a chiral polyhydroxyketone with high diasteroselctivity.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuequan Lu et al.
Organic letters, 7(8), 1645-1648 (2005-04-09)
[reaction: see text] A new analogue of (2S,3R)-ceramide (2) with a methylene group at C4 has been synthesized from d-tartaric acid (3) by using Tebbe methylenation as the key step. Compound 2 exhibited markedly higher antiproliferative activity on mouse embryonic
Mauricio A Cuellar et al.
Bioorganic & medicinal chemistry, 11(12), 2489-2497 (2003-05-22)
The Diels-Alder reaction between two polygodial-derived dienes and simple quinones to yield substituted naphtho- and anthraquinones, is described. The in vitro trypanocide activity for the series was determined. Two of the new compounds showed an activity ten and two times
Transformation of carbonyl to vinylidene groups in the pi-conjugated peripheral substituent of chlorophyll derivatives by Tebbe reagent
Tamiaki H, et al.
Tetrahedron Letters, 57(7), 788-790 (2016)
Ketone Methylenation Using the Tebbe and Wittig Reagents-A Comparison
Pine SH, et al.
Synthesis, 1991(2), 165-167 (1991)
Stereoselective synthesis of C-glycosides via Tebbe methylenation and Claisen rearrangement
Godage HY and Fairbanks AJ
Tetrahedron Letters, 41(39), 7589-7593 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service