Skip to Content
Merck

364975

Sigma-Aldrich

(Triphenylphosphine)copper hydride hexamer

90%

Synonym(s):

Cuprous hydride triphenylphosphine hexamer, Hydrido(triphenylphosphine)copper(I) hexamer, Stryker’s reagent, Triphenylphosphine–Copper(I) hydride Hexamer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3PCuH]6
CAS Number:
Molecular Weight:
1961.04
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Assay

90%

reaction suitability

reagent type: reductant

functional group

phosphine

SMILES string

[CuH].[CuH].[CuH].[CuH].[CuH].[CuH].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12.c%13ccc(cc%13)P(c%14ccccc%14)c%15ccccc%15.c%16ccc(cc%16)P(c%17ccccc%17)c%18ccccc%18

InChI

1S/6C18H15P.6Cu.6H/c6*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;;;;;;;;;;/h6*1-15H;;;;;;;;;;;;

InChI key

PYHLOCIDGQNZFY-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
199982223662370193
reaction suitability

reagent type: reductant

reaction suitability

-

reaction suitability

-

reaction suitability

-

General description

Triphenylgermanium hydride is a hydrogen donor.[1]

Application

Catalyst for:
  • Conjugate reduction of ortho-substituted cinnamic esters to form copper enolates
  • Chemoselective preparation of alcohols via hydrogenation of aldehydes
  • 1,2-addition/transmetalation reactions
  • Synthesis of monomer for preparation of phosphorus- and silicon-containing epoxy resins
  • Hydrosilylation reactions
  • Chiral hydrogenation reactions
  • Hydrostannation of activated alkynes
Some useful synthetic applications of (Triphenylphosphine)copperhydride hexamer include stoichiometric and catalytic hydrogenations, chemoselective conjugate reductions, as well as regiospecific and stereoselectiveconjugate hydride reductions.[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium (II)-catalyzed isomerization of olefins with tributyltin hydride.
Kim IS, et al.
The Journal of Organic Chemistry, 72(14), 5424-5426 (2007)
Aldrichimica Acta, 23, 50-50 (1990)
Elliot L Bennett et al.
Inorganic chemistry, 53(6), 2963-2967 (2014-02-28)
A structural, spectroscopic, and computational investigation of Stryker's reagent, [HCu{P(C6H5)3}]6, and its isotopomers has provided new insights into the complex. Neutron diffraction shows that the hydrides are best described as edge bridging rather than face bridging. The combination of infrared
Elliot Bennett et al.
Inorganic chemistry, 54(5), 2213-2220 (2015-02-12)
CuH is a material that appears in a wide diversity of circumstances ranging from catalysis to electrochemistry to organic synthesis. There are both aqueous and nonaqueous synthetic routes to CuH, each of which apparently leads to a different product. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service