Recommended Products
Quality Level
Assay
99%
mp
>300 °C (lit.)
functional group
amide
SMILES string
CC(=O)NC1=NC(=O)NC=C1
InChI
1S/C6H7N3O2/c1-4(10)8-5-2-3-7-6(11)9-5/h2-3H,1H3,(H2,7,8,9,10,11)
InChI key
IJCKBIINTQEGLY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Four stable conformers of N4-acetylcytosine, and the one of the stable form containing an intramoleuclar six-membered ring have been reported by theoretical calculations.The infrared and FT-Raman spectra of N4-acetylcytosine in the solid phase has been reported.
Application
N4-Acetylcytosine may be used in the preparation of cytidine. It may be used in the preparation of 1-(4-azido-4-deoxy-β-D-glucopyranosyl)cytosine.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and properties of N-(2, 3, 5-tri-O-acetyl-D-ribofuranosyl) maleimide.
The Journal of Organic Chemistry, 40(1), 24-28 (1975)
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 112, 139-145 (2013-05-15)
The infrared and FT-Raman spectra of N4-acetylcytosine in the solid phase have been recorded, and the molecular geometrical parameters were optimized using B3LYP and MP2 methods with 6-311++G(d,p) basis set. The theoretical calculations indicate the presence of four stable conformers
Nucleosides. 63. Synthetic studies on nucleoside antibiotics. 3. Total synthesis of 1-(4-amino-4-deoxy-beta-d-glucopyranosyluronic acid)cytosine, the nucleoside moiety of gougerotin.
The Journal of organic chemistry, 35(1), 231-236 (1970-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service