Skip to Content
Merck
All Photos(4)

Key Documents

224294

Sigma-Aldrich

Thiophosphoryl chloride

98%

Synonym(s):

Phosphorus sulfochloride, Phosphorus thiochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PSCl3
CAS Number:
Molecular Weight:
169.40
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.555 (lit.)

bp

125 °C (lit.)

mp

−35 °C (lit.)

solubility

benzene: soluble(lit.)
carbon disulfide: soluble(lit.)
carbon tetrachloride: soluble(lit.)
chloroform: soluble(lit.)

density

1.668 g/mL at 25 °C (lit.)

SMILES string

ClP(Cl)(Cl)=S

InChI

1S/Cl3PS/c1-4(2,3)5

InChI key

WQYSXVGEZYESBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Thiophosphoryl chloride was used in the synthesis of O-ethyl dichlorothiophosphate.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Coloşi-Esca et al.
Journal of applied toxicology : JAT, 4(5), 230-235 (1984-10-01)
The rat acute oral LD50 value of PSCl3 is 750 mg per kg body weight. A rat 90-day oral feeding study at dose levels of 7.5 mg (P1) and 37.5 mg (P5) per kg resulted in increased body weight gain.
Mohammed D Zidan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 58(8), 1577-1582 (2002-08-09)
A new route has been devised, leading to the production of phosphorus thiotrihalides, SPX3 where X = F, Br and I by an on-line process using phosphorus thiotrichloride, SPCl3 as starting compound gassed over the following heated salts NaF, KBr
Sarah E Lee et al.
Chembiochem : a European journal of chemical biology, 12(4), 633-640 (2011-02-10)
The elucidation of signalling pathways relies heavily upon the identification of protein kinase substrates. Recent investigations have demonstrated the efficacy of chemical genetics using ATP analogues and modified protein kinases for specific substrate labelling. Here we combine N(6) -(cyclohexyl)ATPγS with
J T Slama et al.
Analytical biochemistry, 209(1), 143-149 (1993-02-15)
Elemental [35S]sulfur was shown to equilibrate with the sulfur of thiophosphoryl chloride when these materials are heated together. This isotopic exchange reaction is the basis of a convenient, microscale synthesis of high specific activity [35S]PSCl3. [35S]Thiophosphoryl chloride is otherwise not
[Hygienic substantiation of the maximum permissible level of phosphorus thiotrichloride in reservoir water].
V M Tomkiv
Gigiena i sanitariia, (11)(11), 68-70 (1988-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service