Recommended Products
Quality Level
Assay
98%
bp
256-260 °C (lit.)
mp
129-131 °C (lit.)
SMILES string
CC(C)c1ncc[nH]1
InChI
1S/C6H10N2/c1-5(2)6-7-3-4-8-6/h3-5H,1-2H3,(H,7,8)
InChI key
FUOZJYASZOSONT-UHFFFAOYSA-N
General description
2-Isopropylimidazole is an heterocyclic building block. The nitration of 2-isopropylimidazole has been reported.
Application
2-Isopropylimidazole may be used in the preparation of following compounds:
- Tetragonal nickel(II) complexes, NiL4X2 (X = Cl-, Br-, I-, SCN-, SeCN-, NO3-, ClO4-, PF6- or NiL4XClO4 (X = Cl-, Br-, I-, SCN-).
- 1-Dodecyl-2-isopropylimidazole.
- Bromidotetrakis(2-isopropyl-1H-imidazole-[κN3)copper(II) bromide.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
FEBS letters, 579(10), 2065-2070 (2005-04-07)
To identify a novel class of antibiotics, we have developed a high-throughput genetic system for targeting the homodimerization (HD system) of histidine kinase (HK), which is essential for a bacterial signal transduction system (two-component system, TCS). By using the HD
Tetragonal nickel (II) complexes with singlet and triplet ground states: Complexes with 2-isopropyl imidazole.
J. Inorg. Nucl. Chem., 38(10), 1891-1896 (1976)
Acta crystallographica. Section E, Structure reports online, 67(Pt 10), m1338-m1338 (2011-11-09)
The Cu(II) atom in the title salt, [CuBr(C(6)H(10)N(2))(4)]Br, is coordinated in a square-pyramidal geometry by four imidazole N atoms and one bromide anion that is located at the apex of the pyramid. The cations and the anions form a two-dimensional
Nitration Studies on 2-Isopropylimidazole.
Canadian Journal of Chemistry, 50(21), 3472-3477 (1972)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service