All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C4H4ClN3O2
CAS Number:
Molecular Weight:
161.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
148-150 °C (lit.)
functional group
chloro
nitro
SMILES string
Cn1cnc(c1Cl)[N+]([O-])=O
InChI
1S/C4H4ClN3O2/c1-7-2-6-4(3(7)5)8(9)10/h2H,1H3
InChI key
OSJUNMSWBBOTQU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
5-Chloro-1-methyl-4-nitroimidazole is an 4-nitroimidazole derivative.
Application
5-Chloro-1-methyl-4-nitroimidazole (CMNI) is suitable for use in the rapid mix experiments to investigate the mechanism of anomalous radiosensitization of mammalian cells by CMNI. It may be used in the synthesis of 5-aryl-1-methyl-4-nitroimidazoles, via Suzuki coupling with arylboronic acids, catalyzed by dichlorobis-(triphenylphosphine)palladium(II), K2CO3 and tetrabutylammonium bromide.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J Watras et al.
Neoplasma, 34(3), 253-259 (1987-01-01)
The transplantable rhabdomyosarcoma in WAG/Rij rats was used to test the in vivo effectiveness of 1-methyl-2-chloro-4-nitroimidazole (P13) and its analog 1-(2-hydroxy-3-methoxy-propyl)-2-chloro-4-nitroimidazole (P40) as tumor-cell radiosensitizers after their i.p. administration at low doses. The results indicate that both compounds administered repeatedly
Are ortho-substituted 4-nitroimidazoles a new generation of radiation-induced arylating agents?
E D Clarke et al.
International journal of radiation biology and related studies in physics, chemistry, and medicine, 37(4), 463-468 (1980-04-01)
Rapid-mix studies on the anomalous radiosensitization of mammalian cells by 5-chloro-1-methyl-4-nitromidazole.
M E Watts et al.
International journal of radiation biology and related studies in physics, chemistry, and medicine, 38(6), 673-675 (1980-12-01)
The kinetics of the reaction of 'anomalous' 4-nitroimidazole radiosensitizers with thiols.
P Wardman
International journal of radiation biology and related studies in physics, chemistry, and medicine, 41(2), 231-235 (1982-02-01)
Occupational allergic contact dermatitis from 5-chloro-1-methyl-4-nitroimidazole.
R Jolanki et al.
Contact dermatitis, 36(1), 53-54 (1997-01-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service