About This Item
Recommended Products
Assay
98%
form
solid
mp
287 °C (dec.) (lit.)
functional group
nitro
SMILES string
[O-][N+](=O)c1ncc[nH]1
InChI
1S/C3H3N3O2/c7-6(8)3-4-1-2-5-3/h1-2H,(H,4,5)
InChI key
YZEUHQHUFTYLPH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- tert-butyl 2-(2-nitro-1H-imidazol-1-yl)ethylcarbamate
- 1-(2-(tert-butyldimethylsilyloxy)ethyl)-2-nitro-1H-imidazole
- 2-fluoro-N-(2-(2-nitro-1H-imidazol-1-yl)ethyl)acetamide
- 2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethyl 2-fluoroacetate
- radiolabeling precursors - the bromo substituted analogs
- nitroimidazole substituted boronic acids, precursors for imaging hypoxic tissue
- potential site-selective radiosensitizers for estrogen receptor-rich tumors
Packaging
Analysis Note
50 mg/mL in NH4OH : soluble, clear, green-yellow, to very dark green-yellow to very dark yellow
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service