Skip to Content
Merck
All Photos(1)

Key Documents

366412

Sigma-Aldrich

N-Benzyl-N-methylethanolamine

technical grade, 90%

Synonym(s):

2-(N-Benzyl-N-methylamino)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
165.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

95-105 °C/2 mmHg (lit.)

density

1.017 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl
phenyl

SMILES string

CN(CCO)Cc1ccccc1

InChI

1S/C10H15NO/c1-11(7-8-12)9-10-5-3-2-4-6-10/h2-6,12H,7-9H2,1H3

InChI key

WOUANPHGFPAJCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

N-Benzyl-N-methylethanolamine is a tertiary benzylamine. Electrochemical alkoxylation of N-benzyl-N-methylethanolamine is reported. Rate of proton exchange of N-benzyl-N-methylethanolamine in aqueous HCl has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proton exchange mechanisms of some tertiary benzylamines.
Leyden DE and Morgan WR.
The Journal of Physical Chemistry, 73(9), 2924-2929 (1969)
The Anodic Oxidation of Organic Compounds. II. The Electrochemical Alkoxylation of Tertiary Amines.
Weinberg NL and Brown EA.
The Journal of Organic Chemistry, 31(12), 4058-4061 (1966)
Jessica Paul et al.
Journal of chromatography. A, 1366, 38-44 (2014-10-02)
In 2000 the implementation of quality by design (QbD) was introduced by the Food and Drug Administration (FDA) and described in the ICH Q8, Q9 and Q10 guidelines. Since that time, systematic optimization strategies for purification of biopharmaceuticals have gained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service