Merck
  • Optimization of a preparative multimodal ion exchange step for purification of a potential malaria vaccine.
Optimization of a preparative multimodal ion exchange step for purification of a potential malaria vaccine.
Journal of chromatography. A (2014-10-02)
Jessica Paul, Sonja Jensen, Arthur Dukart, Gesine Cornelissen
ABSTRACT
In 2000 the implementation of quality by design (QbD) was introduced by the Food and Drug Administration (FDA) and described in the ICH Q8, Q9 and Q10 guidelines. Since that time, systematic optimization strategies for purification of biopharmaceuticals have gained a more important role in industrial process development. In this investigation, the optimization strategy was carried out by adopting design of experiments (DoE) in small scale experiments. A combination method comprising a desalting and a multimodal ion exchange step was used for the experimental runs via the chromatographic system ÄKTA™ avant. The multimodal resin Capto™ adhere was investigated as an alternative to conventional ion exchange and hydrophobic interaction resins for the intermediate purification of the potential malaria vaccine D1M1. The ligands, used in multimodal chromatography, interact with the target molecule in different ways. The multimodal functionality includes the binding of proteins in spite of the ionic strength of the loading material. The target protein binds at specific salt conditions and can be eluted by a step gradient decreasing the pH value and reducing the ionic strength. It is possible to achieve a maximized purity and recovery of the product because degradation products and other contaminants do not bind at specific salt concentrations at which the product still binds to the ligands.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets