357839
1,5,6,7-Tetrahydro-4H-indol-4-one
98%
Synonym(s):
4,5,6,7-Tetrahydro-4-oxoindole, NSC 131681
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
98%
mp
188-190 °C (lit.)
functional group
ketone
SMILES string
O=C1CCCc2[nH]ccc12
InChI
1S/C8H9NO/c10-8-3-1-2-7-6(8)4-5-9-7/h4-5,9H,1-3H2
InChI key
KASJZXHXXNEULX-UHFFFAOYSA-N
General description
Iodination of 1,5,6,7-tetrahydro-4H-indol-4-one using 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) yields α-iodo derivative as the main product.
Application
1,5,6,7-Tetrahydro-4H-indol-4-one may be used in the preparation of:
- 4-oxo-4,5,6,7-tetrahydro-1H-indole-2-carbonitrile
- methyl 2-(4-oxo-4,5,6,7-tetrahydro-1H-indol-1-yl)acrylate
- 3-(4-oxo-4,5,6,7-tetrahydro-1H-indol-1-yl)propanenitrile
- potent and orally active 5-HT1A agonists, (R)-(+)- and (S)-(-)-1-formyl-6,7,8,9-tetrahydro-N,N-dipropyl-3H-benz[e]indol-8-amines
- Reactant in synthesis of psammopemmin A as antitumor agent
- Reactant in synthesis of a 1,3,4,5-tetrahydrobenzindole β-ketoesters and tricyclic tetrahydrobenzindoles via C-H insertion reactions
- Reactant in preparation of tricyclic indole and dihydroindole derivatives as inhibitors of guanylate cyclase
- Reactant in preparation of condensed pyrroloindoles via Pd-catalyzed intramolecular C-H bond functionalization of (halobenzyl)pyrroles
- Reactant in enantioselective preparation of arylalkenyl indoles via asymmetric C-H insertion of rhodium carbenoids followed by Cope rearrangement-elimination
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of (R)-and (S)-1-formyl-6, 7, 8, 9-tetrahydro-N, N-(dipropyl)-3H-benz [e] indol-8-amines: potent and orally active 5-HT1A receptor agonists.
Journal of Heterocyclic Chemistry, 31(1), 129-139 (1994)
Synthesis of pyrrolo [1, 2-a] indole-1, 8 (5H)-diones as new synthons for developing novel tricyclic compounds of pharmaceutical interest.
ARKIVOC (Gainesville, FL, United States), 5, 97-106 (2004)
Direct α-iodination of aryl alkyl ketones by elemental iodine activated by 1-chloromethyl-4-fluoro-1, 4-diazoniabicyclo [2.2. 2] octane bis (tetrafluoroborate).
Synthesis, 06, 0853-0858 (2003)
DNA binders: 1. evaluation of DNA-interactive ability, design, and synthesis of novel intercalating agents
Letters in Drug Design & Discovery, 6(1), 56-62 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service