Skip to Content
Merck
All Photos(1)

Key Documents

357642

Sigma-Aldrich

4′-tert-Butylacetophenone

97%

Synonym(s):

1-[4-(1,1-Dimethylethyl)phenyl]ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4COCH3
CAS Number:
Molecular Weight:
176.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.52 (lit.)

bp

107-108 °C/5 mmHg (lit.)

mp

17-18 °C (dimorphic) (lit.)

density

0.964 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1ccc(cc1)C(C)(C)C

InChI

1S/C12H16O/c1-9(13)10-5-7-11(8-6-10)12(2,3)4/h5-8H,1-4H3

InChI key

UYFJYGWNYQCHOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4′-tert-Butylacetophenone is a para-substituted acetophenone.

Application

4′-tert-Butylacetophenone may be used in the synthesis of 2-pyridone derivatives and their fluorescence spectra has been evaluated.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some 3-cyano-4, 6-diaryl-2-pyridones with luminiscent properties.
Kuthan J, et al.
Collection of Czechoslovak Chemical Communications, 44(8), 2409-2416 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service