Skip to Content
Merck
All Photos(3)

Key Documents

349089

Sigma-Aldrich

2-Cyanobenzaldehyde

98%

Synonym(s):

2-Formylbenzonitrile, Phthalaldehydonitrile (8CI), o-Cyanobenzaldehyde, o-Formylbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4CHO
CAS Number:
Molecular Weight:
131.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

103-105 °C (lit.)

functional group

aldehyde

SMILES string

O=Cc1ccccc1C#N

InChI

1S/C8H5NO/c9-5-7-3-1-2-4-8(7)6-10/h1-4,6H

InChI key

QVTPWONEVZJCCS-UHFFFAOYSA-N

General description

Aldol addition of enolizable 1,3-dicarbonyl compounds to 2-cyanobenzaldehyde in the presence of a tertiary amine has been reported. Biotransformation of 2-cyanobenzaldehyde by Euglena gracilis Z cultured photohetero-trophically has been reported.

Application

2-Cyanobenzaldehyde may be used:
  • in the base-catalyzed one-pot synthesis of 3-substituted isoindolinones
  • in the synthesis of 3-oxo-2,3-dihydro-1H-isoindoles, via Baylis-Hillman reaction
  • in the synthesis of 3-(N-substituted amino)-1-isoindolenones

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of baylis-hillman methodology in a new synthesis of 3-oxo-2, 3-dihydro-1H-isoindoles.
Song YS, et al.
Journal of Heterocyclic Chemistry, 40(5), 939-942 (2003)
The aldol addition of readily enolizable 1, 3-dicarbonyl compounds to 2-cyanobenzaldehyde in the synthesis of novel 3-substituted isoindolinones.
More V, et al.
Synthesis, 2011(18), 3027-3031 (2011)
Novel synthesis of 3-(N-substituted amino)-1-isoindolenones from 2-cyanobenzaldehyde with amines.
Sato R, et al.
Chemistry Letters (Jpn), 9, 1599-1602 (1984)
Marcus Angelin et al.
The Journal of organic chemistry, 75(17), 5882-5887 (2010-08-10)
The mechanism of a base-catalyzed one-pot reaction of 2-cyanobenzaldehyde and primary nitroalkanes, to produce 3-substituted isoindolinones, has been investigated. A route starting with a nitroaldol (Henry) reaction, followed by a subsequent cyclization and rearrangement, was supported by intermediate analogue synthesis
Biotransformation of aromatic aldehydes and related compounds by Euglena gracilis Z.
Noma Y, et al.
Phytochemistry, 30(9), 2969-2972 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service