Skip to Content
Merck
All Photos(1)

Key Documents

346128

Sigma-Aldrich

1-(Trifluoroacetyl)imidazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F3N2O
CAS Number:
Molecular Weight:
164.09
Beilstein:
608904
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

137 °C (lit.)
45-46 °C/14 mmHg (lit.)

density

1.442 g/mL at 25 °C (lit.)

functional group

fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)n1ccnc1

InChI

1S/C5H3F3N2O/c6-5(7,8)4(11)10-2-1-9-3-10/h1-3H

InChI key

SINBGNJPYWNUQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(Trifluoroacetyl)imidazole (N-(Trifluroroacetyl)imidazole) is an effective reagent for specific trifluoroacetylation of an aminomethyl side chain of certain nucleosides.

Application

1-(Trifluoroacetyl)imidazole may be employed as derivatization reagent for the detection of mustard gas degradation products, thiodiglycol and thiodiglycol sulfoxide using gas chromatography-tandem mass spectrometry. It may be used as reagent for the trifluoroacetylation of amines. It was used for derivatization of ethanolamine phospholipid-alkenal Michael adducts for GC-MS analysis. It was used in the determination of quinolinic acid, a kynurenine metabolite, by GC/MS.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandrine Bacot et al.
Journal of lipid research, 48(4), 816-825 (2007-01-16)
Hydroxy-alkenals, such as 4-hydroxy-2(E)-nonenal (4-HNE; from n-6 fatty acids), are degradation products of fatty acid hydroperoxides, including those generated by free radical attack of membrane polyunsaturated fatty acyl moieties. The cytotoxic effects of hydroxy-alkenals are well known and are mainly
Direct microwave promoted trifluoroacetylation of aromatic amines with trifluoroacetic acid.
Salazar J, et al.
Journal of Fluorine Chemistry, 124(1), 111-113 (2003)
Stanisław Popiel et al.
Analytical chemistry, 86(12), 5865-5872 (2014-05-17)
A method for detecting mustard gas degradation products thiodiglycol (TDG) and thiodiglycol sulfoxide (TDGO) in water and sediment samples using gas chromatography-tandem mass spectrometry (GC-MS/MS) after derivatization with 1-(trifluoroacetyl)imidazole (TFAI) was described. Selected reaction monitoring mode (SRM) of tandem mass
A Chiarugi et al.
FEBS letters, 453(1-2), 197-200 (1999-07-14)
The rabbit lens has an elevated content of 3-hydroxykynurenine (30HKYN) in spite of a very low activity of the enzymes leading to its synthesis. The iris/ciliary body, on the contrary, has very high activity of 30HKYN synthesizing enzymes but a
Trifluoroacetylation in Organic Synthesis: Reagents, Developments and Applications in the Construction of Trifluoromethylated Compounds.
Lopez SE, et al.
Current Organic Synthesis, 7(5), 414-432 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service