Skip to Content
Merck
All Photos(1)

Key Documents

290920

Sigma-Aldrich

Ethyl pentafluoropropionate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2F5CO2C2H5
CAS Number:
Molecular Weight:
192.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.301 (lit.)

bp

75-76 °C (lit.)

density

1.299 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)C(F)(F)C(F)(F)F

InChI

1S/C5H5F5O2/c1-2-12-3(11)4(6,7)5(8,9)10/h2H2,1H3

InChI key

DBOFMRQAMAZKQY-UHFFFAOYSA-N

Application

Ethyl pentafluoropropionate has been used:
  • as pentafluoroethyl source in the direct synthesis of pentafluoroethyl copper (CuC2F5)
  • in the synthesis of C-6 substituted fluoroalkenyl 2,4-dimethoxypyrimidine derivative

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and structural characterization of the C-6 fluoroalkylated pyrimidine derivatives.
Kristafor S, et al.
Journal of Molecular Structure, 923(1), 19-23 (2009)
Hiroki Serizawa et al.
Organic letters, 16(13), 3456-3459 (2014-06-14)
The direct synthesis of pentafluoroethyl copper (CuC2F5) from a cuprate reagent and ethyl pentafluoropropionate as one of the most economical and useful pentafluoroethyl sources was accomplished. The advantages of this method are; all the reagents employed are low-cost and operationally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service