Skip to Content
Merck
All Photos(2)

Key Documents

345474

Sigma-Aldrich

Lithium trimethylsilanolate

95%

Synonym(s):

Trimethylsilanol lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOLi
CAS Number:
Molecular Weight:
96.13
Beilstein:
3686900
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

SMILES string

[Li+].C[Si](C)(C)[O-]

InChI

1S/C3H9OSi.Li/c1-5(2,3)4;/h1-3H3;/q-1;+1

InChI key

OXOZHAWWRPCVGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Enhanced process and composition control for atomic layer deposition with lithium trimethylsilanolate: This study reports on the novel lithium precursor, lithium trimethylsilanolate (LiTMSO), for atomic layer deposition, comparing it with other lithium precursors for improved process and composition control (Ruud et al., 2017).

  • Trimethylsiloxy based metal complexes as electrolyte additives for high voltage application in lithium ion cells: The paper discusses the synthesis of lithium trimethylsilanolate and its use as an additive to enhance the performance of high voltage lithium ion cells (Imholt et al., 2017).

  • Application of trimethylsilanolate alkali salts in organic synthesis: This review covers various applications of lithium trimethylsilanolate in organic synthesis, highlighting its role in different reactions (Bürglová and Hlaváč, 2018).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grzegorz Piechota et al.
Bioresource technology, 103(1), 16-20 (2011-10-29)
The removal and determination of trimethylsilanol (TMSOH) in landfill gas has been studied before and after the special E3000-ITC System. The system works according to principle of temperature swing. The performance of TMSOH and humidity removal was 20% and more
D M Hui et al.
Clinica chimica acta; international journal of clinical chemistry, 302(1-2), 171-188 (2000-11-14)
We developed a new assay method for fluoride anion (F(-)) a specific metabolite of sarin. Trimethyifluorosilane (TMFS) was derivatized from F(-) with trimethylsilanol, and TMFS was detected with a GC-flame ionization detector (FID) and capillary column system. The linear range
A Isquith et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(3), 263-266 (1988-03-01)
Six organosilicon compounds which had been found to have clastogenic activity in an in vitro battery of genotoxicity assays were evaluated in rat bone marrow cytogenetic assays for assessing clastogenicity in an in vivo system. None of the six compounds
Todd A Duncombe et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13765-13770 (2012-09-01)
We introduce the wetting barrier ratchet, a digital microfluidic technology for directed drop transport in an open air environment. Cyclic drop footprint oscillations initiated by orthogonal vibrations as low as 37 μm in amplitude at 82 Hz are rectified into
Diane M Coe et al.
Organic & biomolecular chemistry, 1(7), 1106-1111 (2003-08-21)
A convenient and efficient method for the cleavage of 1,3-oxazolidin-5-ones and 1,3-oxazolidin-2-ones utilising potassium trimethylsilanolate in tetrahydrofuran is described. The benzyloxycarbonyl-protecting group is readily removed under the reaction conditions, whereas the N-benzoyl group is stable. A synthesis of (R)-salmeterol exploiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service