Merck
Removal and determination of trimethylsilanol from the landfill gas.
Bioresource technology (2011-10-29)
Grzegorz Piechota, Manfred Hagmann, Roman Buczkowski
ABSTRACT
The removal and determination of trimethylsilanol (TMSOH) in landfill gas has been studied before and after the special E3000-ITC System. The system works according to principle of temperature swing. The performance of TMSOH and humidity removal was 20% and more than 90%, respectively. The six of active carbons and impinger method were tested on the full-scale landfill in Poland for TMSOH and siloxanes determination. The extraction method and absorption in acetone were used. The concentration of TMSOH and siloxanes were found in range from 23.6 to 29.2 mg/m3 and from 18.0 to 38.9 mg/m3, respectively. The content of TMSOH in biogas originating from landfill was 41% out of all siloxanes. Moreover, the used system is alternative to other existing technique of landfill gas purification.
Sigma-Aldrich
Trimethylsilanol, ≥97.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium trimethylsilanolate, 95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Potassium trimethylsilanolate, 90%, technical grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Lithium trimethylsilanolate, 95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Tris(trimethylsilyl) phosphite, ≥95.0% (GC)
Pricing and availability is not currently available.